Assay Details
| Gene Symbol: UROC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165974.1 | NP_001159446.1 | 19 - 20 | 2025 | 71 | |
| NM_144639.2 | NP_653240.1 | 18 - 19 | 1845 | 71 | ||
| GenBank mRNA | AB075869.1 | - | 19 - 20 | 2005 | 71 | |
| AK055862.1 | - | 18 - 19 | 1823 | 71 | ||
| BC115405.1 | - | 18 - 19 | 1823 | 40076084 | 71 | |
| BC115406.1 | - | 18 - 19 | 1823 | 40076086 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001741.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031637.hg.1 | TC03001741.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031637.hg.1 | ENST00000290868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031637.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031637.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031637.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012282.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | ENST00000290868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | ENST00000383579.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | NM_001165974.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | NM_144639.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300191899.hg.1 | TC0300012282.hg.1 | Clariom D TC |

UROC1
Genomic Map