Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039796.hg.1 |
TC02002509.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039796.hg.1 |
ENST00000263816.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039796.hg.1 |
NM_004525.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039795.hg.1 |
TC02002509.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039795.hg.1 |
ENST00000263816.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039795.hg.1 |
ENST00000443831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02039795.hg.1 |
NM_004525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208438.hg.1 |
ENST00000263816.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208438.hg.1 |
NM_004525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208438.hg.1 |
ENST00000263816.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208438.hg.1 |
NM_004525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208438.hg.1 |
TC0200014851.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
ENST00000263816.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
ENST00000443831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
NM_004525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
ENST00000263816.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
ENST00000443831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
NM_004525.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200208437.hg.1 |
TC0200014851.hg.1 |
Clariom D TC |