Assay Details
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018073.7 | NP_060543.5 | 2 - 3 | 722 | 86 | |
GenBank mRNA | AF360739.1 | - | 2 - 3 | 675 | 86 | |
AF439153.1 | - | 1 - 2 | 432 | 86 | ||
AK056523.1 | - | 2 - 3 | 684 | 86 | ||
AK291166.1 | - | 2 - 3 | 634 | 86 | ||
AK310888.1 | - | 2 - 3 | 631 | 86 | ||
AL832730.1 | - | 2 - 3 | 672 | 86 | ||
AY005802.1 | - | 1 - 2 | 108 | 86 | ||
BC075058.2 | - | 1 - 2 | 456 | 30915367 | 86 | |
BC109063.2 | - | 2 - 3 | 641 | 40033795 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
