Assay Details
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304496.1 | NP_001291425.1 | 2 - 3 | 330 | 71 | |
NM_018073.7 | NP_060543.5 | 3 - 4 | 813 | 71 | ||
GenBank mRNA | AF360739.1 | - | 3 - 4 | 766 | 71 | |
AF439153.1 | - | 2 - 3 | 523 | 71 | ||
AK056523.1 | - | 3 - 4 | 775 | 71 | ||
AK291166.1 | - | 3 - 4 | 725 | 71 | ||
AK301120.1 | - | 2 - 3 | 213 | 71 | ||
AK310888.1 | - | 3 - 4 | 722 | 71 | ||
AL832730.1 | - | 3 - 4 | 763 | 71 | ||
BC075058.2 | - | 2 - 3 | 547 | 30915367 | 71 | |
BC109063.2 | - | 3 - 4 | 732 | 40033795 | 71 | |
DA833140.1 | - | 2 - 3 | 213 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001311.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK301120.1,NM_001304496.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11016675.hg.1 | ENST00000300747.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11016675.hg.1 | ENST00000544055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11016675.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11016675.hg.1 | TC11001311.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174752.hg.1 | TC1100009884.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
