Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005402.hg.1 |
ENST00000278456.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005402.hg.1 |
ENST00000418331.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005402.hg.1 |
NM_002843.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11005402.hg.1 |
TC11000412.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
ENST00000418331.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
ENST00000613246.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
ENST00000615445.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
NM_002843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
ENST00000418331.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
ENST00000613246.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
ENST00000615445.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
NM_002843.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100153497.hg.1 |
TC1100007507.hg.1 |
Clariom D TC |