Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006050.hg.1 |
TC02000402.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006050.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006050.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
ENST00000478888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007907.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154607.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154607.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154607.hg.1 |
ENST00000377938.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200154607.hg.1 |
NM_019617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200154607.hg.1 |
TC0200007907.hg.1 |
Clariom D TC |