Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032141.3 | NP_115517.1 | 3 - 4 | 210 | 92 | |
| XM_017025210.1 | XP_016880699.1 | 3 - 4 | 1040 | 92 | ||
| XM_017025211.1 | XP_016880700.1 | 4 - 5 | 238 | 92 | ||
| GenBank mRNA | AF161358.1 | - | 3 - 4 | 189 | 92 | |
| AK313963.1 | - | 3 - 4 | 197 | 92 | ||
| AL136806.1 | - | 3 - 4 | 201 | 92 | ||
| BC016313.1 | - | 4 - 5 | 224 | 92 | ||
| BC040118.1 | - | 3 - 4 | 190 | 5176197 | 92 | |
| BC046116.2 | - | 3 - 4 | 212 | 92 | ||
| BC062425.1 | - | 3 - 4 | 202 | 92 | ||
| BC105044.1 | - | 3 - 4 | 197 | 8144047 | 92 | |
| BC105046.1 | - | 3 - 4 | 197 | 8144049 | 92 | |
| CR533555.1 | - | 3 - 4 | 173 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC016313.1, XM_017025210.1, XM_017025211.1 transcripts.
Set Membership: |
|

NSRP1
Genomic Map