Assay Details
| Gene Symbol: CCT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006430.3 | NP_006421.2 | 3 - 4 | 436 | 191 | |
| GenBank mRNA | AF026291.1 | - | 3 - 4 | 374 | 191 | |
| AK129860.1 | - | 3 - 4 | 345 | 191 | ||
| AK295330.1 | - | 3 - 4 | 209 | 191 | ||
| AK307856.1 | - | 4 - 5 | 370 | 191 | ||
| AK312586.1 | - | 3 - 4 | 378 | 191 | ||
| AK315975.1 | - | 4 - 5 | 612 | 191 | ||
| BC106933.1 | - | 3 - 4 | 369 | 40033761 | 191 | |
| BC106934.1 | - | 3 - 4 | 372 | 40033762 | 191 | |
| U38846.1 | - | 3 - 4 | 272 | 191 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK295330.1, AK315975.1 transcripts.
Set Membership: |
|

CCT4
Genomic Map