Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
TC01002791.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
ENST00000370855.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
ENST00000370859.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
ENST00000535611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
ENST00000535790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
ENST00000536707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
NM_001130058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043487.hg.1 |
NM_152697.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
ENST00000370855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
ENST00000370859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
NM_001130058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
NM_152697.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
ENST00000370855.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
ENST00000370859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
NM_001130058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
NM_152697.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209071.hg.1 |
TC0100014617.hg.1 |
Clariom D TC |