Assay Details
| Gene Symbol: SPON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006108.3 | NP_006099.2 | 5 - 6 | 945 | 116 | |
| GenBank mRNA | AB018305.2 | - | 4 - 5 | 430 | 116 | |
| AB051390.1 | - | 5 - 6 | 701 | 116 | ||
| AK074803.1 | - | 5 - 6 | 891 | 116 | ||
| AK291780.1 | - | 5 - 6 | 893 | 116 | ||
| BC136513.1 | - | 5 - 6 | 887 | 9020503 | 116 | |
| BC136563.1 | - | 5 - 6 | 887 | 9020553 | 116 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000211.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002841.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002841.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002841.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002841.hg.1 | TC11000211.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148660.hg.1 | TC1100006911.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |

SPON1
Genomic Map