Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021643.3 | NP_067675.1 | 1 - 2 | 1703 | 93 | |
| GenBank mRNA | AK313237.1 | - | 1 - 2 | 753 | 93 | |
| AY245544.1 | - | 1 - 2 | 1686 | 93 | ||
| BC002637.2 | - | 1 - 2 | 753 | 3607549 | 93 | |
| D87119.1 | - | 1 - 2 | 1492 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000082.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
negative regulation of protein kinase activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of MAP kinase activity
negative regulation of interleukin-10 biosynthetic process
negative regulation of fat cell differentiation
nucleotide binding
protein kinase activity
protein kinase inhibitor activity
transcription factor binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
ubiquitin-protein transferase regulator activity
protein phosphorylation
negative regulation of protein kinase activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of MAP kinase activity
negative regulation of interleukin-10 biosynthetic process
negative regulation of fat cell differentiation
nucleotide binding
protein kinase activity
protein kinase inhibitor activity
transcription factor binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
ubiquitin-protein transferase regulator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001050.hg.1 | TC02000082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001050.hg.1 | ENST00000155926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001050.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001050.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001049.hg.1 | TC02000082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001049.hg.1 | ENST00000155926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001049.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001046.hg.1 | TC02000082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001046.hg.1 | ENST00000155926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001046.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001047.hg.1 | TC02000082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001047.hg.1 | ENST00000155926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001047.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001047.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001048.hg.1 | TC02000082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001048.hg.1 | ENST00000155926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001048.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146366.hg.1 | TC0200006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146363.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146363.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146363.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146363.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146363.hg.1 | TC0200006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146365.hg.1 | TC0200006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146362.hg.1 | TC0200006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146364.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146364.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146364.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146364.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146364.hg.1 | TC0200006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200146361.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146361.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146361.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200146361.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200146361.hg.1 | TC0200006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000155926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000381465.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000405331.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | ENST00000483034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NM_021643.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NR_027303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006757.hg.1 | NR_027303.1 | Refseq |

TRIB2
Genomic Map