Assay Details
| Gene Symbol: OAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134939.1 | NP_001128411.1 | 2 - 3 | 339 | 59 | |
| NM_001301371.1 | NP_001288300.1 | 2 - 3 | 226 | 59 | ||
| NM_016178.2 | NP_057262.2 | 2 - 3 | 374 | 59 | ||
| GenBank mRNA | AF175296.1 | - | 2 - 3 | 374 | 59 | |
| AY713300.1 | - | 2 - 3 | 313 | 59 | ||
| BC073949.1 | - | 2 - 3 | 338 | 5416827 | 59 | |
| BI830988.1 | - | 2 - 3 | 226 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

OAZ3
Genomic Map