Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002188.hg.1 |
ENST00000442544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002188.hg.1 |
NM_005215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002188.hg.1 |
TC18000184.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000412726.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000442544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000579702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000581580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
NM_005215.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
TC1800007340.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000412726.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000442544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000579702.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
ENST00000581580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150295.hg.1 |
NM_005215.3 |
Refseq |