Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011921.hg.1 |
ENST00000522077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010374.hg.1 |
ENST00000506459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011921.hg.1 |
ENST00000522077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010374.hg.1 |
ENST00000506459.1 |
Ensembl |