Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
TC01002300.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
ENST00000264203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
ENST00000375142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
ENST00000375144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
ENST00000375145.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
ENST00000401084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
ENST00000433834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035249.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035246.hg.1 |
TC01002300.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035246.hg.1 |
ENST00000264203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035246.hg.1 |
ENST00000375144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035246.hg.1 |
ENST00000401084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035246.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035246.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
TC01002300.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
ENST00000264203.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
ENST00000375142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
ENST00000375144.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
ENST00000401084.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035247.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197248.hg.1 |
TC0100013134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197249.hg.1 |
TC0100013134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000264202.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000433834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000264202.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
ENST00000433834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197253.hg.1 |
TC0100013134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000433834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
ENST00000433834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197252.hg.1 |
TC0100013134.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000433834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000264203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000375142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000375144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
ENST00000433834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_001206540.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_001206541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_001282162.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
NM_004930.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100197251.hg.1 |
TC0100013134.hg.1 |
Clariom D TC |