Assay Details
| Gene Symbol: PRRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006723286.2 | XP_006723349.1 | 1 - 2 | 93 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006723286.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |

PRRG2
Genomic Map