Assay Details
| Gene Symbol: PRRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000951.2 | NP_000942.1 | 1 - 2 | 153 | 105 | |
| NM_001316335.1 | NP_001303264.1 | 1 - 2 | 153 | 105 | ||
| XM_006723286.2 | XP_006723349.1 | 2 - 3 | 156 | 105 | ||
| GenBank mRNA | BC026032.1 | - | 1 - 2 | 110 | 3960215 | 105 |
| CD013954.1 | - | 1 - 2 | 135 | 105 | ||
| CD609900.1 | - | 1 - 2 | 160 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000740.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC026032.1,NM_000951.2,NM_001316335.1,XM_006723286.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19010345.hg.1 | ENST00000246794.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010345.hg.1 | ENST00000543867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010345.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010345.hg.1 | TC19000740.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010343.hg.1 | ENST00000246794.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010343.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010343.hg.1 | TC19000740.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010344.hg.1 | ENST00000246794.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010344.hg.1 | ENST00000543867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010344.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19010344.hg.1 | TC19000740.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900166209.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166209.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166209.hg.1 | TC1900008546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166209.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166209.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | TC1900008546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166205.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008546.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | TC1900008546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166204.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | TC1900008546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | ENST00000596700.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166203.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | TC1900008546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000246794.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000596877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | ENST00000597121.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166207.hg.1 | NM_000951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900166208.hg.1 | ENST00000543867.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900166208.hg.1 | TC1900008546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900166208.hg.1 | ENST00000543867.2 | Ensembl |

PRRG2
Genomic Map