Assay Details
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016246.2 | NP_057330.2 | 4 - 5 | 544 | 63 | |
XM_005258969.3 | XP_005259026.1 | 4 - 5 | 553 | 63 | ||
GenBank mRNA | AF126781.1 | - | 4 - 5 | 390 | 63 | |
AY358430.1 | - | 4 - 5 | 383 | 63 | ||
BC006283.2 | - | 4 - 5 | 337 | 4026397 | 63 | |
BC006294.1 | - | 4 - 5 | 345 | 4025809 | 63 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001699.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023160.hg.1 | ENST00000263278.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023160.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023160.hg.1 | TC19001699.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023161.hg.1 | ENST00000263278.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023161.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023161.hg.1 | TC19001699.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023158.hg.1 | ENST00000263278.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023158.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023158.hg.1 | TC19001699.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023163.hg.1 | ENST00000263278.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023163.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023163.hg.1 | TC19001699.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023159.hg.1 | ENST00000263278.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023159.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023159.hg.1 | TC19001699.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19023162.hg.1 | ENST00000263278.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19023162.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19023162.hg.1 | TC19001699.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192868.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192863.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192866.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000596349.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011099.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192867.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | ENST00000599157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192861.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | ENST00000595764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192860.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192869.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192869.hg.1 | NM_016246.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900192869.hg.1 | TC1900011099.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900192869.hg.1 | ENST00000263278.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900192869.hg.1 | NM_016246.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 116588 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 228351 | - | - |
