Assay Details
Gene Symbol: GUCA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000409.4 | NP_000400.2 | 4 - 5 | 1030 | 84 | |
NM_001319061.1 | NP_001305990.1 | 4 - 5 | 1401 | 84 | ||
NM_001319062.1 | NP_001305991.1 | 3 - 4 | 854 | 84 | ||
XM_006715073.3 | XP_006715136.1 | 3 - 4 | 243 | 84 | ||
XM_011514537.2 | XP_011512839.1 | 4 - 5 | 1291 | 84 | ||
XM_011514539.1 | XP_011512841.1 | 4 - 5 | 893 | 84 | ||
GenBank mRNA | AK125780.1 | - | 4 - 5 | 1332 | 84 | |
BC031663.1 | - | 4 - 5 | 971 | 5172128 | 84 | |
L36859.1 | - | 2 - 3 | 379 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000580.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
visual perception
phototransduction
regulation of rhodopsin mediated signaling pathway
positive regulation of guanylate cyclase activity
cellular response to calcium ion
calcium ion binding
calcium sensitive guanylate cyclase activator activity
guanylate cyclase regulator activity
signal transduction
visual perception
phototransduction
regulation of rhodopsin mediated signaling pathway
positive regulation of guanylate cyclase activity
cellular response to calcium ion
calcium ion binding
calcium sensitive guanylate cyclase activator activity
guanylate cyclase regulator activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | TC06000580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | ENST00000053469.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | ENST00000372965.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | ENST00000394237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | ENST00000541991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006541.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000623004.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000623004.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | ENST00000623004.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600155734.hg.1 | TC0600008038.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000053469.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000372963.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000394237.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000418175.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000541991.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000623004.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000623004.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | ENST00000623004.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008038.hg.1 | NM_000409.4 | Refseq |
