Assay Details
Gene Symbol: PROZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256134.1 | NP_001243063.1 | 2 - 3 | 146 | 131 | |
XM_017020812.1 | XP_016876301.1 | 3 - 4 | 277 | 131 | ||
XM_017020813.1 | XP_016876302.1 | 2 - 3 | 260 | 131 | ||
GenBank mRNA | M55671.1 | - | 2 - 3 | 146 | 131 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000418.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13004680.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13004680.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004680.hg.1 | TC13000418.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13004679.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13004679.hg.1 | ENST00000375547.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13004679.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004679.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004679.hg.1 | TC13000418.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13004681.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13004681.hg.1 | ENST00000375547.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13004681.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004681.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004681.hg.1 | TC13000418.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154597.hg.1 | TC1300008111.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300154596.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154596.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154596.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154596.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154596.hg.1 | TC1300008111.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | ENST00000493630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008111.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | ENST00000342783.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | ENST00000375547.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | NM_001256134.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | NM_003891.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154595.hg.1 | TC1300008111.hg.1 | Clariom D TC |
