| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009923.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13009923.hg.1 |
TC13000890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165774.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165774.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165774.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165774.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165774.hg.1 |
TC1300009879.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300165773.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165773.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165773.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300165773.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300165773.hg.1 |
TC1300009879.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
ENST00000446789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009879.hg.1 |
NR_034002.1 |
Refseq |