Assay Details
Gene Symbol: CCM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080625.3 | NP_542192.2 | 1 - 2 | 41 | 72 | |
XM_006723706.3 | XP_006723769.1 | 1 - 2 | 104 | 72 | ||
XM_011528566.1 | XP_011526868.1 | 1 - 2 | 104 | 72 | ||
XM_011528569.1 | XP_011526871.1 | 1 - 2 | 104 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000204.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_080625.3,XM_006723706.3,XM_011528566.1,XM_011528569.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
