Assay Details
| Gene Symbol: PJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014819.4 | NP_055634.3 | 1 - 2 | 136 | 155 | |
| GenBank mRNA | AK291759.1 | - | 1 - 2 | 70 | 155 | |
| BM469517.1 | - | 1 - 2 | 156 | 155 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001664.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291759.1,NM_014819.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
long-term memory
regulation of protein kinase A signaling
protein ubiquitination
hippo signaling
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
protein kinase A catalytic subunit binding
protein kinase A regulatory subunit binding
ubiquitin protein ligase activity
long-term memory
regulation of protein kinase A signaling
protein ubiquitination
hippo signaling
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
protein kinase A catalytic subunit binding
protein kinase A regulatory subunit binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05022512.hg.1 | TC05001664.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022512.hg.1 | ENST00000361189.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022512.hg.1 | ENST00000361557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022512.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022512.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022519.hg.1 | TC05001664.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022519.hg.1 | ENST00000361189.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022519.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022518.hg.1 | TC05001664.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022518.hg.1 | ENST00000361189.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022518.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180711.hg.1 | TC0500011672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180716.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180716.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180716.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180716.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180716.hg.1 | TC0500011672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500180717.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180717.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180717.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500180717.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500180717.hg.1 | TC0500011672.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361189.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000361557.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000511624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | ENST00000512822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011672.hg.1 | NM_014819.4 | Refseq |

PJA2
Genomic Map