Assay Details
| Gene Symbol: DOPEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320714.1 | NP_001307643.1 | 16 - 17 | 2961 | 97 | |
| NM_005128.3 | NP_005119.2 | 16 - 17 | 2852 | 97 | ||
| XM_017028509.1 | XP_016883998.1 | 13 - 14 | 2355 | 97 | ||
| GenBank mRNA | AB023150.2 | - | 13 - 14 | 2321 | 97 | |
| AJ237839.2 | - | 16 - 17 | 2858 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21001061.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001273.hg.1 | TC21001061.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001273.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001273.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147517.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147517.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147517.hg.1 | TC2100007039.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147517.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147517.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |

DOPEY2
Genomic Map