Assay Details
Gene Symbol: SQLE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003129.3 | NP_003120.2 | 5 - 6 | 1867 | 117 | |
XM_011517246.2 | XP_011515548.1 | 5 - 6 | 965 | 117 | ||
GenBank mRNA | AF098865.1 | - | 5 - 6 | 1096 | 117 | |
AK225085.1 | - | 5 - 6 | 1157 | 117 | ||
AK295302.1 | - | 5 - 6 | 767 | 117 | ||
AK301792.1 | - | 5 - 6 | 759 | 117 | ||
AK313384.1 | - | 5 - 6 | 985 | 117 | ||
BC017033.1 | - | 5 - 6 | 1099 | 3846346 | 117 | |
BX647400.1 | - | 5 - 6 | 1801 | 117 | ||
D78129.1 | - | 3 - 4 | 385 | 117 | ||
D78130.1 | - | 5 - 6 | 1204 | 117 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000729.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08009908.hg.1 | TC08000729.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009908.hg.1 | ENST00000265896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009908.hg.1 | ENST00000523430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009908.hg.1 | ENST00000541193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009908.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08009909.hg.1 | TC08000729.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009909.hg.1 | ENST00000265896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009909.hg.1 | ENST00000523430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009909.hg.1 | ENST00000541193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009909.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08009907.hg.1 | TC08000729.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08009907.hg.1 | ENST00000265896.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009907.hg.1 | ENST00000523430.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009907.hg.1 | ENST00000541193.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08009907.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000520493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000520493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000520493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160202.hg.1 | TC0800008783.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160204.hg.1 | TC0800008783.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800160200.hg.1 | TC0800008783.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000265896.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000518931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000520493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000520493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000520493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000521232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | ENST00000523430.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008783.hg.1 | NM_003129.3 | Refseq |
