Assay Details
| Gene Symbol: RSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317942.1 | NP_001304871.1 | 2 - 3 | 260 | 97 | |
| NM_178565.4 | NP_848660.3 | 3 - 4 | 939 | 97 | ||
| XM_011517018.1 | XP_011515320.1 | 3 - 4 | 955 | 97 | ||
| XM_011517019.1 | XP_011515321.1 | 3 - 4 | 955 | 97 | ||
| XM_017013395.1 | XP_016868884.1 | 3 - 4 | 928 | 97 | ||
| GenBank mRNA | AK123023.1 | - | 3 - 4 | 902 | 97 | |
| AK123027.1 | - | 1 - 2 | 69 | 97 | ||
| AY358166.1 | - | 3 - 4 | 592 | 97 | ||
| BC027938.1 | - | 2 - 3 | 260 | 5201681 | 97 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001523.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK123027.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
Wnt signaling pathway
positive regulation of Wnt signaling pathway
bone mineralization
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
negative regulation of odontogenesis of dentin-containing tooth
lung growth
epithelial tube branching involved in lung morphogenesis
trachea cartilage morphogenesis
dopaminergic neuron differentiation
positive regulation of canonical Wnt signaling pathway
receptor binding
protein binding
heparin binding
osteoblast differentiation
Wnt signaling pathway
positive regulation of Wnt signaling pathway
bone mineralization
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
negative regulation of odontogenesis of dentin-containing tooth
lung growth
epithelial tube branching involved in lung morphogenesis
trachea cartilage morphogenesis
dopaminergic neuron differentiation
positive regulation of canonical Wnt signaling pathway
receptor binding
protein binding
heparin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019973.hg.1 | TC08001523.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019973.hg.1 | ENST00000276659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019973.hg.1 | ENST00000517939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019973.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019972.hg.1 | TC08001523.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019972.hg.1 | ENST00000276659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019972.hg.1 | ENST00000378439.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019972.hg.1 | ENST00000517781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019972.hg.1 | ENST00000517939.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019972.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177917.hg.1 | TC0800011452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800177922.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177922.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177922.hg.1 | TC0800011452.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000276659.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000517939.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000520026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521502.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000521956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | ENST00000522333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_001282863.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011452.hg.1 | NM_178565.4 | Refseq |

RSPO2
Genomic Map