Assay Details
| Gene Symbol: PIP4K2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005028.4 | NP_005019.2 | 7 - 8 | 1047 | 149 | |
| XM_006717450.2 | XP_006717513.1 | 6 - 7 | 889 | 149 | ||
| XM_011519496.1 | XP_011517798.1 | 7 - 8 | 823 | 149 | ||
| XM_017016330.1 | XP_016871819.1 | 7 - 8 | 881 | 149 | ||
| XM_017016331.1 | XP_016871820.1 | 7 - 8 | 976 | 149 | ||
| XM_017016332.1 | XP_016871821.1 | 6 - 7 | 689 | 149 | ||
| GenBank mRNA | AK294817.1 | - | 7 - 8 | 828 | 149 | |
| AK294867.1 | - | 5 - 6 | 434 | 149 | ||
| BC018034.1 | - | 7 - 8 | 1028 | 4817357 | 149 | |
| S78798.1 | - | 7 - 8 | 1113 | 149 | ||
| U14957.2 | - | 7 - 8 | 821 | 149 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001101.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
regulation of autophagy
regulation of phosphatidylinositol 3-kinase signaling
megakaryocyte development
phosphatidylinositol phosphorylation
positive regulation of autophagosome assembly
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
1-phosphatidylinositol-5-phosphate 4-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol biosynthetic process
regulation of autophagy
regulation of phosphatidylinositol 3-kinase signaling
megakaryocyte development
phosphatidylinositol phosphorylation
positive regulation of autophagosome assembly
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
1-phosphatidylinositol-5-phosphate 4-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10013834.hg.1 | ENST00000323883.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013834.hg.1 | ENST00000376573.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013834.hg.1 | ENST00000545335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013834.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10013834.hg.1 | TC10001101.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000432610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000432610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000432610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168368.hg.1 | TC1000010022.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000168369.hg.1 | TC1000010022.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000323883.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000376573.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000422321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000432610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000432610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000432610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000474335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000545335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | ENST00000604912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010022.hg.1 | NM_005028.4 | Refseq |

PIP4K2A
Genomic Map