Assay Details
| Gene Symbol: FOLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016725.2 | NP_057937.1 | 1 - 2 | 198 | 81 | |
| GenBank mRNA | AK223527.1 | - | 1 - 2 | 52 | 81 | |
| BC002947.2 | - | 1 - 2 | 145 | 3956659 | 81 | |
| J05013.1 | - | 1 - 2 | 199 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000754.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC002947.2,J05013.1,NM_016725.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
heart looping
neural crest cell migration involved in heart formation
cardiac neural crest cell migration involved in outflow tract morphogenesis
ER to Golgi vesicle-mediated transport
receptor-mediated endocytosis
folic acid transport
regulation of transforming growth factor beta receptor signaling pathway
axon regeneration
folic acid metabolic process
COPII vesicle coating
regulation of canonical Wnt signaling pathway
pharyngeal arch artery morphogenesis
anterior neural tube closure
cellular response to folic acid
receptor activity
folic acid binding
drug binding
folic acid transporter activity
methotrexate binding
folic acid receptor activity
heart looping
neural crest cell migration involved in heart formation
cardiac neural crest cell migration involved in outflow tract morphogenesis
ER to Golgi vesicle-mediated transport
receptor-mediated endocytosis
folic acid transport
regulation of transforming growth factor beta receptor signaling pathway
axon regeneration
folic acid metabolic process
COPII vesicle coating
regulation of canonical Wnt signaling pathway
pharyngeal arch artery morphogenesis
anterior neural tube closure
cellular response to folic acid
receptor activity
folic acid binding
drug binding
folic acid transporter activity
methotrexate binding
folic acid receptor activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009791.hg.1 | ENST00000312293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009791.hg.1 | ENST00000393681.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009791.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009791.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009791.hg.1 | TC11000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009789.hg.1 | ENST00000312293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009789.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009789.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009789.hg.1 | TC11000754.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009790.hg.1 | ENST00000312293.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009790.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009790.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009790.hg.1 | TC11000754.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161646.hg.1 | TC1100008340.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161647.hg.1 | TC1100008340.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393676.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | ENST00000393681.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_000802.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008340.hg.1 | NM_016729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | ENST00000312293.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | NM_016724.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | NM_016725.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161645.hg.1 | TC1100008340.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 120122 | - | - |

FOLR1
Genomic Map