Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010806.hg.1 |
ENST00000430323.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010806.hg.1 |
ENST00000525361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010806.hg.1 |
ENST00000533655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010806.hg.1 |
ENST00000533750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010806.hg.1 |
NM_145018.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010806.hg.1 |
TC11000854.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000329143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000524921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000525388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000528189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000528262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000532764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000533655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000533750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
NM_145018.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000329143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000524921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000525388.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000528189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000528262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000532764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000533655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
ENST00000533750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
NM_145018.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202147.hg.1 |
TC1100013075.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000329143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000528262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000532764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000533655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000533750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
NM_145018.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000329143.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000528262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000532764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000533655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
ENST00000533750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
NM_145018.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100202149.hg.1 |
TC1100013075.hg.1 |
Clariom D TC |