Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
TC04001820.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
ENST00000319471.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
ENST00000393528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
ENST00000437304.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
ENST00000448662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
ENST00000498125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
NM_001145671.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
NM_001145672.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
NM_001145673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024912.hg.1 |
NM_003603.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000319454.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000319471.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000393528.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000437304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000448662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000451974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000498125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_001145671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_001145672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_001145673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_003603.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000319454.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000319471.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000393528.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000437304.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000448662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000451974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
ENST00000498125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_001145671.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_001145672.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_001145673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
NM_003603.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400186683.hg.1 |
TC0400012644.hg.1 |
Clariom D TC |