Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004521.hg.1 |
ENST00000283351.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004521.hg.1 |
NM_014939.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004521.hg.1 |
TC18000443.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
ENST00000283351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
ENST00000580104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
ENST00000582539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
NM_014939.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
TC1800008405.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
ENST00000283351.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
ENST00000580104.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
ENST00000582539.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800157295.hg.1 |
NM_014939.3 |
Refseq |