Assay Details
| Gene Symbol: EPG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020964.2 | NP_066015.2 | 34 - 35 | 6046 | 89 | |
| XM_011526122.1 | XP_011524424.1 | 34 - 35 | 6046 | 89 | ||
| XM_017025890.1 | XP_016881379.1 | 34 - 35 | 6028 | 89 | ||
| XM_017025891.1 | XP_016881380.1 | 33 - 34 | 5905 | 89 | ||
| XM_017025892.1 | XP_016881381.1 | 33 - 34 | 6071 | 89 | ||
| XM_017025893.1 | XP_016881382.1 | 26 - 27 | 3918 | 89 | ||
| GenBank mRNA | AK023817.1 | - | 18 - 19 | 2725 | 89 | |
| BC130614.1 | - | 17 - 18 | 2579 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000476.hg.1
Set Membership: |
|

EPG5
Genomic Map