Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000300302.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000344114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000379792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000439977.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000563781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000563911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000564678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000568651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000568814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000570273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
NM_001010989.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
NM_001272103.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
NM_014685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000300302.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000344114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000379792.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000439977.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000563781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000563911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000564678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000568651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000568814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
ENST00000570273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
NM_001010989.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
NM_001272103.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
NM_014685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158351.hg.1 |
TC1600007982.hg.1 |
Clariom D TC |