Assay Details
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142288.1 | NP_001135760.1 | 1 - 2 | 480 | 151 | |
NM_006067.4 | NP_006058.1 | 1 - 2 | 480 | 151 | ||
XM_011522813.2 | XP_011521115.1 | 1 - 2 | 682 | 151 | ||
GenBank mRNA | AF005888.1 | - | 1 - 2 | 349 | 151 | |
AK313977.1 | - | 1 - 2 | 462 | 151 | ||
BC001472.2 | - | 1 - 2 | 432 | 3138396 | 151 | |
BC005886.2 | - | 1 - 2 | 432 | 3546377 | 151 | |
BC007445.1 | - | 1 - 2 | 423 | 3009805 | 151 | |
BC020250.1 | - | 1 - 2 | 432 | 4653699 | 151 | |
BE855452.1 | - | 1 - 2 | 354 | 151 | ||
BP307869.1 | - | 1 - 2 | 485 | 151 | ||
BT007132.1 | - | 1 - 2 | 233 | 151 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001321.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018291.hg.1 | ENST00000253457.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018291.hg.1 | ENST00000435200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018291.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018291.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018291.hg.1 | TC16001321.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16018288.hg.1 | ENST00000253457.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018288.hg.1 | ENST00000435200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018288.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018288.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018288.hg.1 | TC16001321.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR16018289.hg.1 | ENST00000253457.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018289.hg.1 | ENST00000435200.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018289.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018289.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018289.hg.1 | TC16001321.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186313.hg.1 | TC1600011115.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186310.hg.1 | TC1600011115.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186309.hg.1 | TC1600011115.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186308.hg.1 | TC1600011115.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000595980.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000596852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000597291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | ENST00000600807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011115.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | ENST00000253457.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | ENST00000435200.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | NM_001142288.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | NM_006067.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600186311.hg.1 | TC1600011115.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 148027 | - | - |
