Assay Details
| Gene Symbol: IDI2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027708.1 | 2 - 3 | 281 | 84 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000420381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000428780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000434470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000437374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | ENST00000536039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024628.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_024629.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006465.hg.1 | NR_027709.1 | Refseq |

IDI2-AS1
Genomic Map