Assay Details
| Gene Symbol: PARP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006437.3 | NP_006428.2 | 8 - 9 | 990 | 104 | |
| XM_011534931.1 | XP_011533233.1 | 7 - 8 | 1040 | 104 | ||
| XM_011534932.2 | XP_011533234.1 | 8 - 9 | 1207 | 104 | ||
| GenBank mRNA | AF057160.1 | - | 8 - 9 | 936 | 104 | |
| AF158255.1 | - | 8 - 9 | 990 | 104 | ||
| AK304399.1 | - | 8 - 9 | 958 | 104 | ||
| BC017815.1 | - | 8 - 9 | 929 | 104 | ||
| D79999.2 | - | 7 - 8 | 889 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000491.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13005547.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005547.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005547.hg.1 | TC13000491.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156419.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156419.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156419.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300156419.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300156419.hg.1 | TC1300008359.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000381989.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000480576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | ENST00000484989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008359.hg.1 | NM_006437.3 | Refseq |

PARP4
Genomic Map