Assay Details
| Gene Symbol: SETDB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320699.1 | NP_001307628.1 | 4 - 5 | 791 | 74 | |
| NM_031915.2 | NP_114121.2 | 5 - 6 | 1148 | 74 | ||
| GenBank mRNA | AF334407.1 | - | 5 - 6 | 1067 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
heart looping
chromosome segregation
mitotic nuclear division
negative regulation of transcription, DNA-templated
cell division
histone H3-K9 methylation
left/right axis specification
DNA binding
protein binding
zinc ion binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
heart looping
chromosome segregation
mitotic nuclear division
negative regulation of transcription, DNA-templated
cell division
histone H3-K9 methylation
left/right axis specification
DNA binding
protein binding
zinc ion binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000481439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000481439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000481439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002324.hg.1 | ENST00000258672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002324.hg.1 | ENST00000317257.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002324.hg.1 | ENST00000354234.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002324.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002324.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13002324.hg.1 | TC13000198.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000258672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000317257.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000354234.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000481439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000481439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000481439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | ENST00000611815.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_001160308.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007164.hg.1 | NM_031915.2 | Refseq |

SETDB2
Genomic Map