Assay Details
| Gene Symbol: MLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015166.3 | NP_055981.1 | 2 - 3 | 810 | 87 | |
| NM_139202.2 | NP_631941.1 | 2 - 3 | 449 | 87 | ||
| XM_011530678.1 | XP_011528980.1 | 2 - 3 | 571 | 87 | ||
| XM_017028671.1 | XP_016884160.1 | 1 - 2 | 238 | 87 | ||
| GenBank mRNA | AF319633.1 | - | 2 - 3 | 372 | 87 | |
| AK124264.1 | - | 2 - 3 | 811 | 87 | ||
| AK297341.1 | - | 2 - 3 | 272 | 87 | ||
| BC028425.2 | - | 2 - 3 | 409 | 4817175 | 87 | |
| BC070042.1 | - | 2 - 3 | 407 | 5285102 | 87 | |
| CR456460.1 | - | 1 - 2 | 228 | 87 | ||
| D25217.3 | - | 2 - 3 | 486 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000907.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
protein transport
vesicle-mediated transport
positive regulation of intracellular transport
regulation of response to osmotic stress
protein oligomerization
cellular response to cholesterol
caveolin-mediated endocytosis
transporter activity
protein binding
protein transporter activity
protein complex binding
transport
ion transport
protein transport
vesicle-mediated transport
positive regulation of intracellular transport
regulation of response to osmotic stress
protein oligomerization
cellular response to cholesterol
caveolin-mediated endocytosis
transporter activity
protein binding
protein transporter activity
protein complex binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000470008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000470008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000470008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015180.hg.1 | ENST00000395876.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015180.hg.1 | ENST00000450140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015180.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015180.hg.1 | TC22000907.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015181.hg.1 | ENST00000395876.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015181.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015181.hg.1 | TC22000907.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | ENST00000311597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | ENST00000395876.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | ENST00000450140.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | ENST00000535444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | ENST00000538737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015177.hg.1 | TC22000907.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015183.hg.1 | ENST00000395876.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015183.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015183.hg.1 | TC22000907.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | TC2200009142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166066.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000311597.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000442311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000470008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000470008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000470008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | ENST00000483836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_015166.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009142.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166071.hg.1 | TC2200009142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166071.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166071.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166071.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166071.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166069.hg.1 | TC2200009142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166069.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166069.hg.1 | NM_139202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166069.hg.1 | ENST00000395876.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166069.hg.1 | NM_139202.2 | Refseq |

MLC1
Genomic Map