Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007612.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007612.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007612.hg.1 |
TC22000394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007616.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007616.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007616.hg.1 |
TC22000394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007614.hg.1 |
ENST00000361934.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007614.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007614.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007614.hg.1 |
TC22000394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007615.hg.1 |
ENST00000361934.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007615.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007615.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007615.hg.1 |
TC22000394.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000617229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007611.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007611.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007611.hg.1 |
TC22000394.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200154456.hg.1 |
TC2200007662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154456.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154456.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154456.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154456.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154459.hg.1 |
TC2200007662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154459.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154459.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154459.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154459.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154455.hg.1 |
TC2200007662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154455.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154455.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154455.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154455.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154460.hg.1 |
TC2200007662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154460.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154460.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154460.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200154460.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000454366.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000466510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000479645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
ENST00000491863.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007662.hg.1 |
NM_016426.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000617229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
ENST00000623117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009016.hg.1 |
NR_024009.1 |
Refseq |