Assay Details
Gene Symbol: SQRDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271213.1 | NP_001258142.1 | 5 - 6 | 643 | 106 | |
NM_021199.3 | NP_067022.1 | 4 - 5 | 836 | 106 | ||
GenBank mRNA | AF042284.1 | - | 4 - 5 | 526 | 106 | |
AF118085.1 | - | 3 - 4 | 357 | 106 | ||
AF151802.1 | - | 4 - 5 | 520 | 106 | ||
AK130140.1 | - | 5 - 6 | 643 | 106 | ||
AK222463.1 | - | 4 - 5 | 535 | 106 | ||
AK313111.1 | - | 4 - 5 | 515 | 106 | ||
BC016836.1 | - | 4 - 5 | 491 | 3884024 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000364.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15003330.hg.1 | ENST00000260324.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15003330.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15003330.hg.1 | TC15000364.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561493.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000561735.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000563296.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000565997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010737.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182323.hg.1 | TC1500010737.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | ENST00000260324.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | ENST00000566934.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | ENST00000568606.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | NM_001271213.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | NM_021199.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500182324.hg.1 | TC1500010737.hg.1 | Clariom D TC |
