Assay Details
| Gene Symbol: PAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002577.4 | NP_002568.2 | 11 - 12 | 1378 | 89 | |
| XM_011512870.2 | XP_011511172.1 | 11 - 12 | 1101 | 89 | ||
| XM_017006501.1 | XP_016861990.1 | 12 - 13 | 1565 | 89 | ||
| GenBank mRNA | AK291339.1 | - | 11 - 12 | 1262 | 89 | |
| BC069613.1 | - | 11 - 12 | 1091 | 7262289 | 89 | |
| U24153.1 | - | 10 - 11 | 1098 | 89 | ||
| U25975.1 | - | 10 - 11 | 963 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001100.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
protein phosphorylation
negative regulation of protein kinase activity
apoptotic process
signal transduction
viral process
phosphorylation
peptidyl-serine phosphorylation
signal transduction by protein phosphorylation
T cell costimulation
Fc-epsilon receptor signaling pathway
regulation of growth
negative regulation of apoptotic process
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
regulation of defense response to virus by virus
positive regulation of peptidyl-tyrosine phosphorylation
T cell receptor signaling pathway
dendritic spine development
positive regulation of protein tyrosine kinase activity
cellular response to organic cyclic compound
cell-cell adhesion
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
protein tyrosine kinase activator activity
identical protein binding
cadherin binding involved in cell-cell adhesion
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
protein phosphorylation
negative regulation of protein kinase activity
apoptotic process
signal transduction
viral process
phosphorylation
peptidyl-serine phosphorylation
signal transduction by protein phosphorylation
T cell costimulation
Fc-epsilon receptor signaling pathway
regulation of growth
negative regulation of apoptotic process
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
regulation of defense response to virus by virus
positive regulation of peptidyl-tyrosine phosphorylation
T cell receptor signaling pathway
dendritic spine development
positive regulation of protein tyrosine kinase activity
cellular response to organic cyclic compound
cell-cell adhesion
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
protein tyrosine kinase activator activity
identical protein binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03019022.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019022.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019022.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019024.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019024.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019024.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019027.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019027.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019027.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019021.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019021.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019021.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019023.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019023.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019023.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019026.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019026.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019026.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000481344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000481344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | ENST00000481344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019025.hg.1 | TC03001100.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019025.hg.1 | ENST00000327134.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019025.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173087.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173089.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173089.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173089.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173089.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173089.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173090.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173090.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173090.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173090.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173090.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173084.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173085.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173086.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173086.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173086.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173086.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173086.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300173083.hg.1 | TC0300010033.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000327134.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000426668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000481344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000481344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | ENST00000481344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010033.hg.1 | NM_002577.4 | Refseq |

PAK2
Genomic Map