Assay Details
Gene Symbol: PPP1R32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170753.1 | NP_001164224.1 | 6 - 7 | 710 | 60 | |
NM_145017.2 | NP_659454.2 | 6 - 7 | 710 | 60 | ||
GenBank mRNA | AK057333.1 | - | 6 - 7 | 703 | 60 | |
BC043246.1 | - | 6 - 7 | 708 | 5296268 | 60 | |
BC053995.1 | - | 5 - 6 | 607 | 5166383 | 60 | |
JF442033.1 | - | 6 - 7 | 683 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11003456.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11006462.hg.1 | TC11003456.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11006462.hg.1 | ENST00000338608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11006462.hg.1 | ENST00000432063.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11006462.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11006462.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200825.hg.1 | TC1100013020.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
