Assay Details
| Gene Symbol: PPP1R32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170753.1 | NP_001164224.1 | 11 - 12 | 1279 | 76 | |
| NM_145017.2 | NP_659454.2 | 12 - 13 | 1339 | 76 | ||
| GenBank mRNA | AK057333.1 | - | 12 - 13 | 1332 | 76 | |
| BC043246.1 | - | 11 - 12 | 1277 | 5296268 | 76 | |
| BC053995.1 | - | 11 - 12 | 1236 | 5166383 | 76 | |
| JF442033.1 | - | 12 - 13 | 1312 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003456.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | TC11003456.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | ENST00000338608.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | ENST00000366212.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | ENST00000432063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006470.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013020.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000338608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000366212.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000432063.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000535545.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000538185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | ENST00000542951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | NM_001170753.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | NM_145017.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200835.hg.1 | TC1100013020.hg.1 | Clariom D TC |

PPP1R32
Genomic Map