Assay Details
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017841.2 | NP_060311.1 | 2 - 3 | 274 | 92 | |
GenBank mRNA | AK000494.1 | - | 2 - 3 | 274 | 92 | |
BC002331.2 | - | 2 - 3 | 257 | 3535474 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11003455.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11006444.hg.1 | TC11003455.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11006444.hg.1 | ENST00000301761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11006444.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000623232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | ENST00000623232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200800.hg.1 | TC1100013018.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000301761.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000359614.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000534878.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000536250.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000537782.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000542794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000543265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000623232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | ENST00000623232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100013018.hg.1 | NM_017841.2 | Refseq |
