Assay Details
Gene Symbol: PPP1R32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145017.2 | NP_659454.2 | 8 - 9 | 883 | 60 | |
GenBank mRNA | AK057333.1 | - | 8 - 9 | 876 | 60 | |
BC053995.1 | - | 7 - 8 | 780 | 5166383 | 60 | |
JF442033.1 | - | 8 - 9 | 856 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
