Assay Details
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178117.1 | NP_001171588.1 | 1 - 2 | 1088 | 158 | |
| NM_004897.4 | NP_004888.2 | 1 - 2 | 1088 | 158 | ||
| XM_006718078.2 | XP_006718141.1 | 1 - 2 | 2762 | 158 | ||
| XM_017016965.1 | XP_016872454.1 | 1 - 2 | 2762 | 158 | ||
| GenBank mRNA | AB209819.1 | - | 1 - 2 | 651 | 158 | |
| AF046914.1 | - | 1 - 2 | 638 | 158 | ||
| AF084943.1 | - | 1 - 2 | 658 | 158 | ||
| AF084944.1 | - | 1 - 2 | 643 | 158 | ||
| AK304624.1 | - | 2 - 3 | 279 | 158 | ||
| AK312937.1 | - | 1 - 2 | 679 | 158 | ||
| AL050356.1 | - | 1 - 2 | 660 | 159 | ||
| AY358938.1 | - | 1 - 2 | 643 | 158 | ||
| BC032504.1 | - | 1 - 2 | 640 | 5243440 | 158 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000619.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ossification
polyphosphate metabolic process
dephosphorylation
bone mineralization
inositol phosphate metabolic process
acid phosphatase activity
phosphohistidine phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
bisphosphoglycerate 3-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
inositol hexakisphosphate 2-phosphatase activity
ossification
polyphosphate metabolic process
dephosphorylation
bone mineralization
inositol phosphate metabolic process
acid phosphatase activity
phosphohistidine phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
bisphosphoglycerate 3-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
inositol hexakisphosphate 2-phosphatase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006778.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006778.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006778.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006782.hg.1 | ENST00000371994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006782.hg.1 | ENST00000371996.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006782.hg.1 | ENST00000546140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006782.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006782.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006782.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006779.hg.1 | ENST00000371994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006779.hg.1 | ENST00000371996.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006779.hg.1 | ENST00000546140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006779.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006779.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006779.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006781.hg.1 | ENST00000371994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006781.hg.1 | ENST00000371996.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006781.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006781.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006781.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | ENST00000371994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | ENST00000371996.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | ENST00000546140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006784.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000156554.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156554.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156554.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156554.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156554.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156555.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156557.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156560.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 188640 | - | - |

MINPP1
Genomic Map