Assay Details
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178117.1 | NP_001171588.1 | 2 - 3 | 1280 | 108 | |
| GenBank mRNA | AF084944.1 | - | 2 - 3 | 835 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000619.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ossification
polyphosphate metabolic process
dephosphorylation
bone mineralization
inositol phosphate metabolic process
acid phosphatase activity
phosphohistidine phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
bisphosphoglycerate 3-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
inositol hexakisphosphate 2-phosphatase activity
ossification
polyphosphate metabolic process
dephosphorylation
bone mineralization
inositol phosphate metabolic process
acid phosphatase activity
phosphohistidine phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
bisphosphoglycerate 3-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity
inositol hexakisphosphate 2-phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | ENST00000371994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | ENST00000371996.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006791.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006792.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006792.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006792.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006792.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006792.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | ENST00000371994.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | ENST00000371996.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | ENST00000546140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006790.hg.1 | TC10000619.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156567.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156568.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156569.hg.1 | TC1000008351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008351.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000371994.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000371996.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000472891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | ENST00000536010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | NM_001178117.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | NM_001178118.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | NM_004897.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156566.hg.1 | TC1000008351.hg.1 | Clariom D TC |

MINPP1
Genomic Map