Assay Details
| Gene Symbol: DENND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318052.1 | NP_001304981.1 | 15 - 16 | 2969 | 103 | |
| NM_015689.4 | NP_056504.3 | 16 - 17 | 3071 | 103 | ||
| NR_134477.1 | 14 - 15 | 2638 | 103 | |||
| XM_005249977.3 | XP_005250034.1 | 16 - 17 | 3602 | 103 | ||
| XM_011516052.2 | XP_011514354.1 | 16 - 17 | 2949 | 103 | ||
| XM_011516053.2 | XP_011514355.1 | 15 - 16 | 3053 | 103 | ||
| XM_011516054.2 | XP_011514356.1 | 16 - 17 | 2854 | 103 | ||
| XM_017011989.1 | XP_016867478.1 | 15 - 16 | 3055 | 103 | ||
| GenBank mRNA | AB033103.2 | - | 15 - 16 | 2969 | 103 | |
| AK290056.1 | - | 16 - 17 | 2840 | 103 | ||
| AK297959.1 | - | 8 - 9 | 1730 | 103 | ||
| BC049193.1 | - | 12 - 13 | 1439 | 103 | ||
| BC132875.1 | - | 14 - 15 | 2666 | 40146897 | 103 | |
More Information
Set Membership: |
|

DENND2A
Genomic Map