Assay Details
Gene Symbol: SEC14L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014692.1 | NP_055507.1 | 1 - 2 | 132 | 88 | |
GenBank mRNA | AB007880.2 | - | 1 - 2 | 132 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000138.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB007880.2,NM_014692.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16002177.hg.1 | ENST00000251170.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16002177.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16002177.hg.1 | TC16000138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16002178.hg.1 | ENST00000251170.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16002178.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16002178.hg.1 | TC16000138.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600189235.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600189235.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600189235.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600189235.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600189235.hg.1 | TC1600011346.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011346.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | ENST00000251170.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | ENST00000587469.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | NM_014692.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600189237.hg.1 | TC1600011346.hg.1 | Clariom D TC |
