Assay Details
| Gene Symbol: NUDT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012758.2 | NP_001012776.1 | 4 - 5 | 507 | 56 | |
| XM_011509260.2 | XP_011507562.1 | 2 - 3 | 945 | 56 | ||
| XM_011509262.1 | XP_011507564.1 | 4 - 5 | 1155 | 56 | ||
| GenBank mRNA | AK298083.1 | - | 3 - 4 | 647 | 56 | |
| BC046352.1 | - | 2 - 3 | 179 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003111.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01048185.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048185.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048185.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048200.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048200.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048200.hg.1 | ENST00000467151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048200.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048184.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048184.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048184.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048193.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048193.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048193.hg.1 | ENST00000467151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048193.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048197.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048197.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048197.hg.1 | ENST00000467151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048197.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048192.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048192.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048192.hg.1 | ENST00000467151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048192.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048196.hg.1 | TC01003111.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048196.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048196.hg.1 | ENST00000467151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048196.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170107.hg.1 | TC0100009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009746.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170112.hg.1 | TC0100009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170103.hg.1 | TC0100009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170111.hg.1 | TC0100009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000444015.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | ENST00000498192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170104.hg.1 | TC0100009746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | ENST00000334513.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | ENST00000460879.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | ENST00000477878.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | NM_001012758.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100170100.hg.1 | TC0100009746.hg.1 | Clariom D TC |

NUDT17
Genomic Map